Cole-Parmer Essentials Electronic Ice Bucket Ceramic Beads
Representative image only.
Cole-Parmer Item # WW-24120-99
Keep your samples cold, clean, and dry
  • Chemically resistant
  • Can be autoclaved at 134°C
  • 1 year warranty
PRODUCT OPTIONS
₹12,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionElectronic Ice Bucket Ceramic Beads
  • Warranty1 year
1 Year Warranty
Key Features
  • Chemically resistant
  • Can be autoclaved at 134°C
  • 1 year warranty
More About this Item
These Techne N°ICE ceramic-coated beads maintain the temperature and support the sample vessels for as long as necessary. The beads are chemically resistant and if required can be autoclaved at 134°C. As a time saving option, the bucket of beads can be incubated in the fridge overnight and simply inserted into the unit as and when required. Compatible with the 24120-00 Electronic Ice Bucket, sold separately.
These N°ICE ceramic-coated beads maintain the temperature and support the sample vessels for as long as necessary. The beads are chemically resistant and if required can be autoclaved at 134°C. As a time saving option, the bucket of beads can be incubated in the fridge overnight and simply inserted into the unit as and when required. Compatible with the 24120-00 Electronic Ice Bucket, sold separately.