Cole-Parmer CB-200D-IB Electronic Ice Bucket; 100-230 V, 50/60 Hz
Representative image only.
Cole-Parmer Item # WW-24120-00
Keep your samples cold, clean, and dry
  • Waterless—no more wet melting ice or floating sample tubes
  • Ceramic-coated beads maintain a 4°C temperature for the entire working day
  • Holds approximately 60 x 1.5 mL microtubes
  • 3 year warranty
  • Previous part number FNOICE
₹190,000.00INR / Each
Often Bought With...
SPECS & DESCRIPTION
Specifications & Description
  • Bath TypeBead
  • Max Temperature (° C)40
  • Min Temperature (° C)0
  • Power (VAC)100 to 230
  • Power (Hz)50/60
  • DescriptionElectronic Ice Bucket, 100-230 V, 50/60 Hz
  • Warranty3 Years
3 Year WarrantyConformite Europeenne
Key Features
  • Waterless—no more wet melting ice or floating sample tubes
  • Ceramic-coated beads maintain a 4°C temperature for the entire working day
  • Holds approximately 60 x 1.5 mL microtubes
  • 3 year warranty
  • Previous part number FNOICE
More About this Item
Use the electronic ice bucket to keep your tubes clean and dry while incubating at sub-ambient temperatures for long periods of time. A removable bucket is filled with ceramic-coated beads that can maintain a constant temperature and support the sample tubes. To save time, the bucket of beads can be incubated in the refrigerator overnight and inserted into the unit before use. Reusable beads can be autoclaved at 273°F (134°C).
A temperature control ensures all contents in the ice bucket are kept at an even temperature, whether cold or warm. A dark plastic lid helps maintain a constant temperature and protects light sensitive samples. View the current temperature on a five-digit orange LED display in either °C or °F.
Please Note: Effective June 2022, the Techne Electronic Ice Bucket Model number FNOICE has now been rebranded Cole-Parmer under Model number CB-200D-IB. All the same great features and specifications, just with a new and improved look.