Cole-Parmer Deep Basket for 12 L Industrial Fluidized Sand Bath, Steel
Representative image only.
Cole-Parmer Item # WW-01184-72
  • Durable steel mesh construction
  • Safely hold samples away from heating elements
  • Easily retrieve samples from the bath
  • Previous part number 7031659
PRODUCT OPTIONS
₹160,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSteel
  • Compatible WithIndustrial fluidized sand bath 01184-20
  • DescriptionDeep Basket for 12 L Industrial Fluidized Sand Bath, Steel
Key Features
  • Durable steel mesh construction
  • Safely hold samples away from heating elements
  • Easily retrieve samples from the bath
  • Previous part number 7031659
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.