Swift Optical M15B-P Binocular Microscope w/Infinity-Corrected Optics; 40x/100x
Representative image only.
Swift Optical Mfr # M15B-P – Item # WW-48924-70
Infinity-corrected optics give excellent images with enhanced contrast
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification40x/100x/400x/1000x
  • Maximum Magnification1000x
  • Head ConfigurationBinocular
  • Eyepiece Diameter (mm)20
  • Eyepiece Magnification10x
  • Stage Dimensions140 x 140 mm
  • Illuminator(s)Halogen, 20 W
  • Power (VAC)110 to 240
  • DescriptionBinocular Microscope w/Infinity-Corrected Optics; 40x/100x
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
This compound microscope features lenses that improve image clarity — ideal for biological applications. Viewing head has 45° inclined eyepieces with diopter and interpupillary adjustment. Siedentopf type head ensures the tube length remains the same when adjusting interpupillary distance. Feature a 20-watt halogen bulbs and an Abbe condenser.
Microscopes feature coarse and fine focusing adjustments, and large 5-1/2" x 5-1/2" (140 x 140 mm) stage with specimen holder. Objective turret is reverse-mounted giving you easy access to the stage.