Cole-Parmer SHM/ADAPT Adapter for Disposable Homogenizer Probes
Representative image only.
Cole-Parmer Item # WW-04720-08
Make the most of your homogenizer
  • Probes available in different sizes and materials
  • Stand for benchtop use
  • Parts and adapters to expand capabilities
  • Compatible with SHM1 and SHM3
  • Adapts disposable probes to homogenizer
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel (SS)
  • Compatible WithDisposable PC homogenizer probes 04720-07
  • DescriptionAdaptor for Disposable Homogenizer Probes
  • Warranty3 year
3 Year Warranty
Key Features
  • Probes available in different sizes and materials
  • Stand for benchtop use
  • Parts and adapters to expand capabilities
  • Compatible with SHM1 and SHM3
  • Adapts disposable probes to homogenizer
  • 3 year warranty
More About this Item
Stainless steel rotor probes work well with large volumes, and disposable polycarbonate probes reduce cross-contamination (autoclave and reuse up to seven times!). Ideal for tissue homogenisation, cell disruption, emulsions, suspensions, and protein extraction.