Cole-Parmer Cover for Shaking Water Baths, Polycarbonate
Representative image only.
Cole-Parmer Item # WW-12152-99
Choose from these accessories to make your unit more versatile
  • Polycarbonate cover for shaking water baths 12152-00 and -02
  • 1 year warranty
  • Previous part number SBS40/4
PRODUCT OPTIONS
₹60,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Compatible WithShaking water baths 12152-00 and 12152-02
  • DescriptionCover for Shaking Water Baths, Polycarbonate
  • Warranty1 Year
1 Year Warranty
Key Features
  • Polycarbonate cover for shaking water baths 12152-00 and -02
  • 1 year warranty
  • Previous part number SBS40/4
More About this Item
It is normal to have a gap between the edge of the shaking water baths and the polycarbonate cover. The gap prevents chamber pressure to build up and allows excess steam to escape and condensate to fall back into the bath.