Cole-Parmer Digital Colony Counter Optional Magnifier, 1.7x
Representative image only.
Cole-Parmer Item # WW-14212-60
Enhance your digital colony counter
  • Easily view small colonies
  • 1 year warranty
  • Previous part number SC6/1
PRODUCT OPTIONS
₹9,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDigital Colony Counter Optional Magnifier, 1.7x
  • Warranty1 year
1 Year Warranty
Key Features
  • Easily view small colonies
  • 1 year warranty
  • Previous part number SC6/1
More About this Item
Choose from a wide range of replacement parts and accessories to extend the life of your digital colony counter and enhance your viewing capabilities.
This optional screw-in magnifier allows you to easily view small colonies.