Cole-Parmer Brass Cooling Plug for Analog Melting Point Apparatus
Representative image only.
Cole-Parmer Item # WW-03013-66
Accessories for your melting point apparatus
  • Use to rapidly reduce heating block temperature after use
  • Previous part number SMP1/2
PRODUCT OPTIONS
₹4,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialBrass
  • Compatible With03013-60, 03013-61
  • DescriptionBrass Cooling Plug for Analog Melting Point Apparatus
Key Features
  • Use to rapidly reduce heating block temperature after use
  • Previous part number SMP1/2