Cole-Parmer Block Heater Insert, Single Wide, 20 x 2 mL Tubes
Representative image only.
Cole-Parmer Item # WW-36610-52
Interchangeable heating blocks offer excellent thermal conductivity
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Single wide
  • Holds 20, 2-mL tubes
  • 1 year warranty
  • Previous part number SHT1/20
PRODUCT OPTIONS
₹8,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Number Of Blocks1
  • Block Number Of Openings20
  • Block TypeAluminum
  • Block - Tube / Vial Size Accepted (mL)2
  • DescriptionBlock Heater Insert, Single Wide, 20 x 2 mL Tubes
  • Warranty1 Year
1 Year Warranty
Key Features
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
  • Single wide
  • Holds 20, 2-mL tubes
  • 1 year warranty
  • Previous part number SHT1/20
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.