Sticht 9285 Compact Vibrating Reed Tachometer; 7,000 to 11,000 rpm
Representative image only.
Sticht Mfr # 9285 – Item # WW-98806-01
  • 1 year warranty
₹100,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCompact Vibrating Reed Tachometer; 7,000 to 11,000 rpm
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Because these tachometers measure speeds by contact, this type of tachometer is the solution to many speed measuring problems, especially checking speeds of totally enclosed rotating equipment. These tachometers operate on the well known and time tested principle of resonance. The instrument is
simply held against motor, turbine, pump, compressor, or other rotating equipment and the speed is shown by the vibration of a steel reed which is tuned to a certain standard speed. Ruggedly constructed, permanent accuracy, simple to use. The instrument imposes no load and cannot be damaged by overspeeding. Accuracy is 0.5% of rated speed.
Compact Vibrating Reed Tachometers
Smaller in size than the standard tachometers at left, these tachometers have smaller and fewer reeds. Being more compact, they are ideal for tool case or pocket use. Moderately priced, they are the answer to many speed measuring problems.