Sterilisable Packaging Material - Bop Reels, Lavender, 12 Cm Width X 200 M
Representative image only.
Sterilisable Packaging Material - Bop Reels, Lavender, 12 Cm Width X 200 M
| Item: 9470246
Call For Price
Call For Price
Product Description
More About this Item
BOP Reels materials exhibit total and uniform peelability characteristics with minimal fibre lift, irrespective of speed or conditions of opening. The BOP range of products will eliminate reprocessing costs and equally help in reducing nosocomial infection, and by the ease of use ultimately increase patient safety.
Technical Characteristics:
Non Tearing and Shaterless Film: A unique Film specifically formulated to eliminate tearing, a major weakness in standard materials. The resistance to tear is some 10 times greater and virtually prevents any shattering on seals when opening. Paper: The paper used for BOP reels incorporates an adopted surface treatment which not only allows uniform and constant heat sealing, but will also reduce considerably the shedding of fibre when opened.
Indicat
Specifications
  • Width (in)5
  • DescriptionSterilisable Packaging Material - Bop Reels, Lavender, 12 Cm Width X 200 M