This item is now discontinued. We recommend the following as suggested replacements: WW-86292-08
Spill-X 78778 Solvent Spill Treatment/Clean Up Kit
Representative image only.
Spill-X Mfr # 78778 – Item # WW-86292-04
Treat chemical spills in the lab or on the plant floor with a single step!
  • Neutralizes hazardous liquid and chemical spills
  • Quick cleanups
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chemical TreatedSolvent
  • Agent ColorBlack
  • DescriptionSolvent Spill Treatment/Clean Up Kit
  • Warranty1 year
1 Year Warranty
Key Features
  • Neutralizes hazardous liquid and chemical spills
  • Quick cleanups
  • 1 year warranty
More About this Item
These acid-neutralizing spill kits contain sorbents and other supplies which allow you to cleanup spills quickly. The easy-to-use shaker containers help neutralize hazardous liquid and chemical spills.
Order agent refills separately.
Spill-X-S Solvent Adsorbent reduces vapors released during cleanup and elevates flash point of flammable organic spills. Treats spills of many common solvents, reagents, and fuels. Does not treat organic peroxides or hydrazine compounds.