Spill-X 76258 Solvent Spill Kit Agent Refill; 16 lb Pail
Representative image only.
Spill-X Mfr # 76258 – Item # WW-86293-54
Neutralizes spills for safer cleanup and handling
  • Granular base reduces product waste and disposal costs
  • Speeds spill cleanup to limit worker exposure and minimize production downtime
  • Non-toxic, free-flowing dry chemical is environmentally friendly
  • Easy identification of agent with unique color
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Chemical TreatedSolvent
  • Agent ColorBlack
  • ColorBlack
  • DescriptionSolvent Spill Kit Agent Refill; 16 lb Pail
  • Warranty1 year
1 Year Warranty
Key Features
  • Granular base reduces product waste and disposal costs
  • Speeds spill cleanup to limit worker exposure and minimize production downtime
  • Non-toxic, free-flowing dry chemical is environmentally friendly
  • Easy identification of agent with unique color
  • 1 year warranty
More About this Item
These refill agents are available in easy-to-store polypropylene containers. Order to replace specific or multipurpose spill treatment kit containers.
Economy-size pail of control agent covers a 3-gallon (11 L) spill. guide and instructions are included with each pail for start-to-finish cleanup.