Spider mixer support stand for 55 gallon vessel, 24"H x 5/8" dia
Representative image only.
Mfr # SSS-55 – Item # WW-50401-71
₹94,300.00INR / Each
Consider alternative
SPECS & DESCRIPTION
Specifications & Description
  • Shaft Diameter (in)0.625
  • Shaft Diameter (mm)16
  • Shaft Length (in)24
  • Shaft Length (mm)610
  • DescriptionSpider mixer support stand for 55 gallon vessel, 24\"H x 5/8\" dia
More About this Item
Spider support stands clamp onto the edges of square or rectangular pails, drums, and tanks. Model 50401-70 fits 5-gallon vessels; model 50401-71 fits 55-gallon vessels with rims up to 5/8" wide. Aluminum base has a stainless steel support rod.