Cole-Parmer VapLock™ Waste Funnel, 4" dia., 1/2" NPT(M), HDPE / PP; 1/ea
Representative image only.
Cole-Parmer Item # WW-12020-66
Safely and securely capture waste and hazardous solvents
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite® Centura safety containers
PRODUCT OPTIONS
₹48,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDPE / PP
  • Compatible WithPorts with 1/2\" NPT(F)
  • Diameter (in)4
  • Diameter (cm)10.2
  • DescriptionWaste Funnel, 4\" dia., 1/2\" NPT(M), HDPE / PP; 1/ea
Key Features
  • Reduces fumes and harmful vapor exhaust
  • Easily connect to Justrite® Centura safety containers
More About this Item
Use these 4" diameter funnels to safely dispose of liquid laboratory waste. A locking hinged lid minimizes leaks and spills when the funnel is closed. Quick-disconnect fitting on models 12020-64 and -65 allows fast and easy connection to Justrite Centura safety containers. Funnels are constructed of high-density polyethylene (HDPE) and polypropylene (PP) with a Viton® gasket.