Spex VapLock™ Threaded Adapter Fitting, 9/16" Hex, Straight, Black Polypropylene, 1/2" NPT(M) x 1/4" NPT(F)
Representative image only.
Spex Item # WW-12020-80
Designed for secure leak-resistant connections
  • 9/16" hex body makes it easy to wrench tighten adapters
  • Thread to thread adapters provide a tight seal
  • Constructed of durable black polypropylene
PRODUCT OPTIONS
₹400.00INR / Each
Idex Wrench Open End, 1/2
₹4,300.00INR / Each
Often Bought With
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialBlack Polypropylene
  • Number Of Ports2
  • Connection Type1/2\" NPT(M) x 1/4\" NPT(F)
  • Thread size NPT(M) (port 1) (in)1/2
  • Thread size NPT(F) (port 2) (in)1/4
  • Flow PatternStraight
  • ColorBlack
  • DescriptionThreaded Adapter Fitting, 9/16\" Hex, Straight, Black Polypropylene, 1/2\" NPT(M) x 1/4\" NPT(F)
Key Features
  • 9/16" hex body makes it easy to wrench tighten adapters
  • Thread to thread adapters provide a tight seal
  • Constructed of durable black polypropylene
More About this Item
These Spex® Vaplock™ threaded adapters allow for quick and easy connection of different thread sizes. Chromatography adapter fittings are ideal for use in Vaplock manifolds, solvent delivery and waste collection, and general chromatography applications.