Cole-Parmer VapLock™ Solvent Waste Kit, PTFE, panel mount, 1x1/2"NPTF, 3x1/4"NPTF, 6x1/4-28UNFF; 1/ea
Representative image only.
Cole-Parmer Item # WW-12019-78
Collect effluent from multiple sources while minimizing harmful solvent exhaust
  • Various cap sizes available to fit your solvent container
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap / Stopper SizePanel mount
  • DescriptionSolvent Waste Kit, PTFE, panel mount, 1x1/2\"NPTF, 3x1/4\"NPTF, 6x1/4-28UNFF; 1/ea
Includes
10-port PTFE manifold with panel mount connection: (6) 1/4"-28 UNF(F), (3) 1/4" NPT(F), and (1) 1/2" NPT(F)Exhaust filter with Breakthrough color indicator PP filter adapter: 1/4" NPTM x 1/2" NPT(M) PP plugs: (3) 1/4" NPT(M) and (1) 1/2" NPT(M) PTFE plugs: (6) 1/4"-28 UNF(M) PP adapters for semi-rigid tubing: (2) 2.2 mm OD, (1) 2.5 mm OD, (2) 1/16" OD, and (3) 1/8" OD PP barbed adapters with 1/4" NPT(M) connection for soft tubing: (1) 1/8" ID, (1) 3/16" ID, (3) 1/4" ID, and (1) 3/8" ID PP barbed Y connector: 1/4" ID 5 feet of 1/8" OD rigid extension tubing with union connections
Key Features
  • Various cap sizes available to fit your solvent container
More About this Item
Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers. Waste kits include a manifold, exhaust filter, adapters, plugs, and tubing—everything you need to install on a typical HPLC instrument. Manifold allows solvent waste to be collected from multiple effluent streams. Exhaust filter minimize exposure to harmful chemical vapors. Fittings and tubing provide leak-tight connections minimizing spills and leaks.
Panel-mount caps fit a 1-1/4" ID opening with panel thickness from 1/16" to 1/2". A 1-3/4" diameter area around the opening is required be free from obstruction for use. Manifold drains to a 1/2" NPT common port base and come with PTFE locking nut.