Cole-Parmer VapLock™ Reversible Control Valve, 2 x 1/4" NPT(M), 0.1-0.25 psi crack pressure, PP & FFKM; 1ea
Representative image only.
Cole-Parmer Item # WW-12020-60
Maintain pressure equilibrium within the bottle for safe solvent handling
  • Valves feature 0.1 PSI breaking pressure
PRODUCT OPTIONS
₹7,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/4\" NPT(M) x 1/4\" NPT(M)
  • Breaking Pressure (PSI)0.1
  • Seal MaterialFFKM
  • Housing MaterialPP
  • Wetted MaterialsPP/FFKM
  • DescriptionReversible Valve, 2 x 1/4\" NPT(M), 0.1-0.25 psi crack pressure, PP & FFKM; 1ea
Key Features
  • Valves feature 0.1 PSI breaking pressure
More About this Item
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Reversible valves serve either purpose depending on the orientation of the valve.