Cole-Parmer VapLock™ Relief Valve, 1/4-28 UNF(M), 0.1-0.25 psi crack pressure, PTFE & Viton; 1ea
Representative image only.
Cole-Parmer Item # WW-12020-54
Maintain pressure equilibrium within the bottle for safe solvent handling
  • Choice of 0.1 to 0.25 psi or 1 to 2.5 psi cracking pressure
PRODUCT OPTIONS
₹5,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/4\"-28 UNF(M)
  • Breaking Pressure (PSI)0.1
  • Seal MaterialViton®
  • Housing MaterialPTFE
  • Wetted MaterialsPTFE/Viton®
  • DescriptionRelief Valve, 1/4-28 UNF(M), 0.1-0.25 psi crack pressure, PTFE & Viton; 1ea
Key Features
  • Choice of 0.1 to 0.25 psi or 1 to 2.5 psi cracking pressure
More About this Item
Valves minimize evaporation and relieve pressure to ensure safe delivery and containment of solvents. Relief valves let pressurized bottle contents escape above the cracking pressure.