Cole-Parmer VapLock™ Port Plug, Polypropylene, 1/2" NPT(M); 10/PK
Representative image only.
Cole-Parmer Item # WW-12020-45
Designed for secure leak-resistant connections
  • Available in multiple materials and sizes
  • Wrap with PTFE tape for leak-resistant connections
PRODUCT OPTIONS
₹1,700.00INR / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeNPT(M)
  • Thread size NPT(M) (port 1) (in)1/2
  • ColorGray
  • DescriptionPort Plug, 1/2\" NPT(M), Polypropylene; 10/pk
Key Features
  • Available in multiple materials and sizes
  • Wrap with PTFE tape for leak-resistant connections
More About this Item
Use plugs to seal unused manifold and bottle cap ports for a secure leak-resistant system. Options for NPT ports are 1/8", 1/4", 3/8", and 1/2" NPT. To ensure a leak-resistant connection, the plugs should be wrapped three to four times with PTFE tape.