Cole-Parmer VapLock™ Filter Adapter, 1/4" NPT(M) x 1/2" NPT(M), polypropylene; 1/ea
Representative image only.
Cole-Parmer Item # WW-12019-87
Various adapter configurations allow quick and easy connection to emission sources
  • Choose from polypropylene or nylon construction
PRODUCT OPTIONS
₹210.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Connection TypeNPT(M) x NPT(M)
  • Thread size NPT(M) (port 1) (in)1/2
  • Thread size NPT(M) (port 2) (in)1/4
  • ColorGray
  • DescriptionFilter Adapter, 1/4\" NPT(M) x 1/2\" NPT(M), polypropylene; 1/ea
Key Features
  • Choose from polypropylene or nylon construction
More About this Item
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. These adapters are NPT(M) x NPT(M), including options for different sizes and choices between straight or elbow.