Cole-Parmer VapLock™ Exhaust Filter with Indicator, 75 g activated carbon, 1/4" NPT(F); 1/ea
Representative image only.
Cole-Parmer Item # WW-12019-80
Reduce harmful chemical emissions and odors from solvent containers
  • Use with indicators to set a replacement schedule
PRODUCT OPTIONS
₹8,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter Media75 g activated carbon with indicator
  • Number of Cartridges1
  • Inlet/Outlet NPT1/4\" NPT(F)
  • DescriptionExhaust Filter with Indicator, 75 g activated carbon, 1/4\" NPT(F); 1/ea
Key Features
  • Use with indicators to set a replacement schedule
More About this Item
These exhaust filters contain activated media to capture harmful chemicals from escaping into the lab. Filters are made of high-density polyethylene and have a 1/4" NPT(F) port. Use the breakthrough indicator with exhaust filters to detect end-of-service life. Color change occurs when the indicator is exposed to organic solvent vapors. Various adapter configurations are available for a quick and easy connection to emission sources.