Cole-Parmer VapLock™ Complete Solvent Collection System, quick disconnect, PTFE manifold, 20 L
Representative image only.
Cole-Parmer Item # WW-12020-70
Complete solution to safely collect solvent waste from laboratory instruments
  • FM-approved container offers added safety
  • Collect solvent waste from up to 10 effluent streams
  • Minimize exposure to harmful chemical vapors
PRODUCT OPTIONS
₹288,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (L)20
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap / Stopper SizeQuick disconnect
  • Bottle ColorTransluscent
  • DescriptionComplete Solvent Collection System, quick disconnect, PTFE manifold, 20 L
FM Approved
Includes
FM-approved 20-L translucent PE container, PTFE manifold, SS quick-disconnect fittings, plugs for any unused ports, and an exhaust filter with color indicator.
Key Features
  • FM-approved container offers added safety
  • Collect solvent waste from up to 10 effluent streams
  • Minimize exposure to harmful chemical vapors
More About this Item
Solvent waste systems come complete with everything you need to safely collect solvent waste from any system that generates solvent waste. Closed-system design reduces harmful and flammable solvent vapor emissions into the environment as well as leaks and spills.
Systems come with an FM-approved, OSHA, NFPA, and EPA compliant 20-L polyethylene (PE) safety container with quick-disconnect connections plus multiple safety features. A pressure relief lid automatically vents between 3 and 5 psi (0.2 and 0.35 bar) reducing any risk of fire, and a stainless steel arrester prevents flashback ignition. An integral valve prevents vapor release and leaks. A large, self-closing opening and an easy-carry handle with fixed-position latch keeps lid open for convenient pouring. Select from PP or PTFE manifold with quick disconnect fitting to connect up to 10 waste lines.