Cole-Parmer VapLock™ Adapters for Exhaust Filters, Justrite® Centura Quick-Disconnect, Straight, Adjustable, Stainless Steel/PTFE/Black Acetal
Representative image only.
Cole-Parmer Item # WW-12019-99
Various adapter configurations allow quick and easy connection to emission sources
  • Suitable for use with Justrite® Centura safety containers
PRODUCT OPTIONS
₹11,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialSS/PTFE/acetal
  • O-Ring MaterialPTFE
  • Connection TypeNPT(M) x Quick Disconnect, adjustable
  • Thread size NPT(M) (port 2) (in)1/4
  • ColorGray/black
  • DescriptionAdjustable Filter Adapter, Justrite Centura, SS / PTFE / black acetal; 1/ea
Key Features
  • Suitable for use with Justrite® Centura safety containers
More About this Item
VapLock exhaust filters have a 1/4" NPT(F) port and may require an adapter to work with your container. Quick-disconnect adapters fit Justrite® Centura safety containers.