Spex VapLock™ 2-Way Isolation Valve, Normally Closed, 0.032" Orifice, PTFE Body, PTFE Diaphragm, 20 psi, 19 µL Internal Volume, with Mounting Holes, 12 VDC
Representative image only.
Spex Mfr # 075T2NC12-32M – Item # WW-12021-45
Fully isolated solenoid allows use with aggressive and high-purity fluids
  • Designed for over 20 million continuous cycles
  • Compact design—ideal for tight spaces
  • Fast response time
  • Very low dead space
PRODUCT OPTIONS
₹32,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/4”-28 UNF ports
  • Orifice (in)0.032
  • Orifice (mm)0.8128
  • Cv0.011
  • Housing MaterialPTFE
  • Max Pressure (PSI)20
  • Power (VDC)12
  • Wetted MaterialsPTFE
  • Flow Pattern2-way, normally closed
  • Description2-Way Isolation Valve, Normally Closed, 0.032\" Orifice, PTFE Body, PTFE Diaphragm, 20 psi, 19 µL Internal Volume, with Mounting Holes, 12 VDC
Key Features
  • Designed for over 20 million continuous cycles
  • Compact design—ideal for tight spaces
  • Fast response time
  • Very low dead space
More About this Item
Easily dispense precise, repeatable volume using solenoid operated isolation valves. Valves feature a diaphragm that separates the flow path from the operating mechanism ensuring an inert fluid path. Specifically designed to fit in tight spaces, compact valves are great for placing within instruments. Small overall size helps keep the dead volume to a minimum. Fast response time of less than 20 ms ensures flow can be turned on or off in an instant.