Cole-Parmer Spex® 3616ST Polished Steel Pellets, 35 mm; 2/Pk
Representative image only.
Cole-Parmer Item # WW-04577-37
Optimize performance of your Cole-Parmer® hydraulic laboratory pellet press
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more
  • For use with evacuable pellet die set 04576-14
  • Ideal for routine pressing of non-abrasive materials
  • Constructed of durable polished steel
PRODUCT OPTIONS
₹87,800.00INR / Pkg of 2
OFTEN BOUGHT WITH…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolished Steel
  • Compatible With04577-46 and 04577-47 press; 04576-14 die set
  • Diameter (mm)35
  • DescriptionPolished Steel Pellets, 35 mm; 2/Pk
Key Features
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more
  • For use with evacuable pellet die set 04576-14
  • Ideal for routine pressing of non-abrasive materials
  • Constructed of durable polished steel
More About this Item
Ensure top results from your Cole-Parmer® hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components. The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.