Cole-Parmer Spex® 1688 MiniG® Homogenizer Foam Block, 2 x 75 mL PC Jars 04575-72; 1/EA
Representative image only.
Cole-Parmer Item # WW-04575-39
Accommodates a full range of sample vials with an adjustable clamp
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Rugged foam block holds two 75 mL polycarbonate jars
PRODUCT OPTIONS
₹4,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsMiniG® Homogenizer
  • MaterialFoam
  • DescriptionMiniG® Homogenizer Foam Block, 2 x 75 mL PC Jars 04575-72; 1/EA
Key Features
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Rugged foam block holds two 75 mL polycarbonate jars
More About this Item
Multiple inserts are available for a variety of homogenizing applications.
Compatible with polycarbonate jars 04575-72.