Cole-Parmer Spex® 2052 Genomax Adjustable Clamp for Titer Plates
Representative image only.
Cole-Parmer Item # WW-41057-00
Quick and efficient extraction of nucleic acids, proteins, and other molecules through an innovative bead beating technique
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Accommodates up to 6 deep well titer plates
  • Quick-locking system allows for easy installation and removal
  • Color indicator to determine when adequate clamping force is applied
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithTemperature Controlled GenoLyte
  • DescriptionGenomax Adjustable Clamp for Titer Plates
Key Features
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Accommodates up to 6 deep well titer plates
  • Quick-locking system allows for easy installation and removal
  • Color indicator to determine when adequate clamping force is applied
More About this Item
Get the accessories you need to properly operate your HG-200 GenoLyte®.
The 2052 clamp accommodates up to 6 deep well titer plates. This new design ensures that titer plates will not leak during homogenization when used with the Spex #2205-50 titer plates and #2206 cap mats. The quick-locking system on the Genomax allows for easy installation and removal of the clamp. Users can load the clamp on the bench top and then lock it into place on the Genomax or leave the clamp in place and load the plates directly into the Genomax.