Cole-Parmer Spex® 1218 GenoLyte® Homogenizer Vial Holder, 1 x 12 mL Vial 04577-64; 1/EA
Representative image only.
Cole-Parmer Item # WW-04575-35
Quick and efficient extraction of nucleic acids, proteins, and other molecules through an innovative bead beating technique
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Holds one 12 mL vial 04577-64 that accepts one 11 mm grinding ball 04500-22
PRODUCT OPTIONS
₹72,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsHG-200 GenoLyte® homogenizer
  • DescriptionGenoLyte® Homogenizer Vial Holder, 1 x 12 mL Vial 04577-64; 1/EA
Key Features
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Holds one 12 mL vial 04577-64 that accepts one 11 mm grinding ball 04500-22
More About this Item
Get the accessories you need to properly operate your HG-200 GenoLyte®.