Cole-Parmer Spex® 2011 Geno/Grinder® Replacement Adjustable Clamp Assembly for HG-600 Geno/Grinder® Homogenizer
Representative image only.
Cole-Parmer Item # WW-04575-41
Adjusts to fit various sizes of titer plates and vials
  • Accommodates up to 6 deep-well titer plates or up to sixteen 50 mL tubes
  • Stack titer plates in the clamp increasing throughput
  • Release mechanism make it easy to slide clamp up and down the shaft
  • Adjustable Clamp Assembly for new model HG-600 Geno/Grinder® 2010
  • Clamp provides 1.25" (3.2 cm) vertical movement
₹225,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGeno/Grinder® Replacement Adjustable Clamp Assembly for HG-600 Geno/Grinder® Homogenizer
Key Features
  • Accommodates up to 6 deep-well titer plates or up to sixteen 50 mL tubes
  • Stack titer plates in the clamp increasing throughput
  • Release mechanism make it easy to slide clamp up and down the shaft
  • Adjustable Clamp Assembly for new model HG-600 Geno/Grinder® 2010
  • Clamp provides 1.25" (3.2 cm) vertical movement
More About this Item
This front loading clamp comes standard in all new HG-600 Geno/Grinder® 2010 but can be purchased separately as a retro-fit.