Cole-Parmer Spex® 3618 Edge Flaring Tool, 31/40 mm; 1/EA
Representative image only.
Cole-Parmer Item # WW-04576-16
Aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques
  • Reinforces sample disks to resist chipping and breaking
  • Makes sample disks easier to remove from the pellet die
  • Minimizes contamination during sample handling
  • Protects the bore of the pellet die from abrasive samples
  • Helps create a tighter fit in the pellet die
  • Edge flaring prevents sample loss and pellet jamming
PRODUCT OPTIONS
₹14,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Diameter (mm)31
  • DescriptionEdge Flaring Tool, 31/40 mm; 1/EA
Key Features
  • Reinforces sample disks to resist chipping and breaking
  • Makes sample disks easier to remove from the pellet die
  • Minimizes contamination during sample handling
  • Protects the bore of the pellet die from abrasive samples
  • Helps create a tighter fit in the pellet die
  • Edge flaring prevents sample loss and pellet jamming
More About this Item
Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
To use, simply press the tapered end of the edge flaring tool into the Spec-Caps® cup.