Cole-Parmer Spex® 2255 Cryogenic Grinding Vial Set; 4/PK
Representative image only.
Cole-Parmer Item # WW-04575-73
Get the most out of your Cole-Parmer® homogenizer
  • Choose from a wide range of accessories for your homogenizer
  • Designed for grinding tough samples such as bone and plant material
  • Vial consists of two end plugs, a polycarbonate cylinder, and two 1/2” (12.7 mm) SS grinding balls
PRODUCT OPTIONS
₹67,600.00INR / Pkg of 4
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsExtractor 04575-79 and Center cylinders 04577-01
  • MaterialPolycarbonate, stainless steel
  • DescriptionCryogenic Grinding Vial Set; 4/PK
Key Features
  • Choose from a wide range of accessories for your homogenizer
  • Designed for grinding tough samples such as bone and plant material
  • Vial consists of two end plugs, a polycarbonate cylinder, and two 1/2” (12.7 mm) SS grinding balls
More About this Item
Optimize your homogenizer with these quality accessories.