Cole-Parmer Spex® 6900XL Cryogenic Gloves, X-Large; 2/PK
Representative image only.
Cole-Parmer Item # WW-04578-06
Protect your hands and arms from ultra-cold temperatures
  • Multilayer insulation yet lightweight and flexible
  • Gloves are sold as a pair
  • Extra-large size
PRODUCT OPTIONS
₹31,600.00INR / Pkg of 2
SPECS & DESCRIPTION
Specifications & Description
  • SizeX-large
  • ColorBlue
  • DescriptionCryogenic Gloves, X-Large; 2/PK
Key Features
  • Multilayer insulation yet lightweight and flexible
  • Gloves are sold as a pair
  • Extra-large size
More About this Item
Wear cryogenic gloves to safely handle materials from the Freezer/Mill® Cryogenic Grinders. Gloves are multilayered to provide maximum protection yet still allow for excellent dexterity and are comfortable to wear for extended periods of time.