Cole-Parmer Spex® HG-100 Cryo-Station Cooling Device Refrigerated by Liquid Nitrogen
Representative image only.
Cole-Parmer Item # WW-04575-81
Hold temperature sensitive samples chilled prior to grinding
  • Fill insulated jacket manually or automatically with liquid nitrogen supply
  • Quickly reach liquid nitrogen temperatures
  • Holds up to two Cryo-Blocks
  • 1 year warranty
₹379,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithCryo-Blocks
  • DescriptionCryo-Station Cooling Device Refrigerated by Liquid Nitrogen
  • Warranty1 year
1 Year Warranty
Key Features
  • Fill insulated jacket manually or automatically with liquid nitrogen supply
  • Quickly reach liquid nitrogen temperatures
  • Holds up to two Cryo-Blocks
  • 1 year warranty
More About this Item
Cryo-Station chamber offers a convenient place to keep samples that must be kept chilled prior to grinding or to preserve pesticides, RNA, or proteins for extraction. The well can hold a chilled cutting board (included) to prepare temperature sensitive samples.