Cole-Parmer Spex® 8019 Multi Vial Adapter, Holds 5 mL Plastic Vials, Aluminum
Representative image only.
Cole-Parmer Item # WW-04576-73
Use adapters to run multiple samples or to hold vial securely in place during grinding
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Simultaneously run seven vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple smaller samples
  • Compatible with 5 mL plastic vials
PRODUCT OPTIONS
₹20,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAluminum
  • Compatible With5 mL plastic grinding vials
  • DescriptionMulti Vial Adapter, Holds 5 mL Plastic Vials, Aluminum
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® Cryogenic Grinders
  • Simultaneously run seven vials in the High-Energy Mixer/Mill® Grinders
  • Excellent for processing multiple smaller samples
  • Compatible with 5 mL plastic vials
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinder with a wide range of adapters and clamps.