Cole-Parmer Spex® 6881 Grinding Vial Set, Mid-Size Vial, 25 mL, PC Cylinder, SS End Plugs and Impactor

Representative image only.
Cole-Parmer Item # WW-61043-08
Accessories for Freezer/Mill® cryogenic grinders to simplify your processes
  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Transparent center to view progress
  • Withstands liquid nitrogen temperatures
  • PC cylinder, SS end plugs and impactor—easy to clean and reusable
  • Sample capacity of 1 to 40 g

PRODUCT OPTIONS

₹78,400.00INR / Pkg of 1

SPECS & DESCRIPTION

Specifications & Description

  • Use with GeneratorsFreezer/Mill® cryogenic grinders 04500-40 to -42 and 04577-92 to -94
  • Min Sample Size (mL)1
  • Min Sample Size (g)3
  • Max Sample Size (mL)25
  • Max Sample Size (g)25
  • MaterialPC cylinder, SS end plugs and impactor
  • DescriptionGrinding Vial Set, Mid-Size Vial, 25 mL, PC Cylinder, SS End Plugs and Impactor

Includes

One set of stainless steel (SS) end plugs, one SS impactor, and four polycarbonate center cylinder.

Key Features

  • Closed grinding vial prevents cross-contamination
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles
  • Vial sets consist of a center cylinder, two end plugs, and an impactor
  • Transparent center to view progress
  • Withstands liquid nitrogen temperatures
  • PC cylinder, SS end plugs and impactor—easy to clean and reusable
  • Sample capacity of 1 to 40 g

More About this Item

Compatible with Freezer/Mill® cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.

The closed grinding vial prevents sample cross-contamination, retains volatiles, and protects hazardous and critical samples. Stainless steel end plugs and impactors are autoclavable and reusable; polycarbonate cylinders are disposable or may be cleaned wit hot water and detergent or RNase/DNase-free solution. Vial set is compatible with CG-400, CG-450, and CG-500 Freezer/Mill® cryogenic grinders. Easily remove end plugs using extractor 04500-50 and mid-size vial adapter 04500-48.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat