Cole-Parmer Spex® 2206 Sealing Mat for Square 96-Well Reinforced Microplate; 1/EA
Representative image only.
Cole-Parmer Item # WW-41010-97
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Square mat specifically designed for the 96-well reinforced titer plate 41010-96
PRODUCT OPTIONS
₹1,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithTiter plate 41010-96
  • DescriptionSealing Mat for Square 96-Well Reinforced Microplate; 1/EA
Key Features
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Square mat specifically designed for the 96-well reinforced titer plate 41010-96
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. 