Cole-Parmer Spex® HG-800D Geno/Grinder® Automated High-Throughput Homogenizer with Clamp for Two Microplates; 115/230

Representative image only.
Cole-Parmer Item # WW-61031-68
Integrates with robotic systems for automated sample processing
  • Continuous operation provides high-throughput processing of samples
  • Fast and efficient extraction of nucleic acids, proteins, and other molecules
  • Accommodates all standard microplates
  • Typical grinding time is two minutes or less
  • Clamp provides 1.25" (3.2 cm) vertical movement
  • Clamp speed from 500 to 1700 strokes/min
  • Clamp holds two microplates
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Height (in)35 7/8
  • Width (in)12 7/8
  • Depth (in)18 7/8
  • Height (cm)91.1
  • Width (cm)32.7
  • Depth (cm)47.9
  • Power (VAC)115 / 230
  • Power (Hz)50/60
  • DescriptionGeno/Grinder® Automated High-Throughput Homogenizer with Clamp for Two Microplates; 115/230
  • Warranty3 Year
3 Year WarrantyConformite EuropeenneMade in the U.S.A.

Includes

Clamp for two microplates and a power cord.

Key Features

  • Continuous operation provides high-throughput processing of samples
  • Fast and efficient extraction of nucleic acids, proteins, and other molecules
  • Accommodates all standard microplates
  • Typical grinding time is two minutes or less
  • Clamp provides 1.25" (3.2 cm) vertical movement
  • Clamp speed from 500 to 1700 strokes/min
  • Clamp holds two microplates
  • 3 year warranty

More About this Item

The HG-800 Geno/Grinder® homogenizer integrates with robotic systems to automate rapid cell disruption of plant and animal tissue through bead beating. This process offers fast and efficient extraction of nucleic acids, proteins, and other molecules. With the average processing time of less than two minutes and the ability to run around the clock, the homogenizer can process a high number of samples.

Easily load and unload microplates—the included clamp syncs with robotic systems to automatically open and close. A unique vigorous vertical grinding motion of the clamp allows increased reproducibility, superior extraction efficiency, and consistency of every sample. Pulverize samples with or without buffer using a variety of grinding media.

The password-protected, touch screen control panel enables you to program run time, rate, cycles, and pause time. Up to 500 protocols can be easily labeled, saved, recalled, and exported via USB.

Effective September 2023, the Spex® SamplePrep 2030 Geno/Grinder® has now been rebranded Cole-Parmer® under model number HG-800 Geno/Grinder®. All the same great features and specifications, just with a new and improved look.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat

Bioz Article and Whitepaper Citations

Powered by Bioz See more details on Bioz