Spex Protects Frequent-Use Hand Soap, Fresh & Clean Scent, 3 oz (90 mL)

Representative image only.
Spex Protects Item # WW-78901-56
Reduce cross-contamination with this laboratory-grade soap
  • Effectively yet gently washes away the most common germs for better hand health
  • Contains natural moisturizers like aloe and lanolin that nourish your skin
  • pH balanced (5.3-5.4) to skin's natural acid mantle barrier
  • Fresh & Clean Scent
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • FormatPurse Size
  • Volume (gallons)0.024
  • Volume (Liters)0.09
  • DescriptionHand Soap, Fresh & Clean Scent, 3 oz (90 mL)
  • Warranty3 year
3 Year Warranty

Key Features

  • Effectively yet gently washes away the most common germs for better hand health
  • Contains natural moisturizers like aloe and lanolin that nourish your skin
  • pH balanced (5.3-5.4) to skin's natural acid mantle barrier
  • Fresh & Clean Scent
  • 3 year warranty

More About this Item

The soothing coconut-based formula with moisturizing lanolin and aloe is gentle to skin, so it cleans without drying or chapping. Used by scientists who wash their hands up to 40 times a day. Available in a 17 oz (500 mL) convenient pump dispenser or 3 oz (90 mL) pocket or purse size.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat