Spectra Por S/P 2 Dialysis Membrane Trial Kit, 12,000-14,000 Dalton, 25mm Flat Width

Representative image only.
Spectra Por Mfr # 132678T – Item # WW-02900-02
Contains everything you needed to evaluate dialysis membranes
  • Ideal for smaller dialysis experiments
  • 1 year warranty

PRODUCT OPTIONS

₹27,800.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Flat Width (mm)25
  • Diameter (mm)16
  • Length Per Pack (ft)16.5
  • Diameter (in)0.63
  • DescriptionDialysis Membrane Trial Kit, 12,000-14,000Dalton 25mm
  • Warranty1 year
1 Year Warranty

Key Features

  • Ideal for smaller dialysis experiments
  • 1 year warranty

More About this Item

These convenient kits contain everything you need for smaller dialysis experiments or evaluating dialysis tubing for a dialysis application. The small roll is ideal for low volume projects. Spectra/Por® 1 through 3 Dialysis Trail Kits are available in a range of nominal molecular weight cut-offs (MWCO) for less specific separations. Spectra/Por grades 1 through 3 are packaged dry with 10% glycerol and contain trace levels of heavy metals and sulfur.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat