Solar Light PMA2132 Visible PAR Sensor 1-13/16"H x 1-9/16" dia
Representative image only.
Solar Light Mfr # PMA2132 – Item # WW-97650-56
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Resolution0.1 µE/s/m2
  • FunctionMeasures light intensity below 0.01 Lux, can be used as a safety detector for hazardous intensities of light
  • Intensity Range20,000 µE/s/m2
  • Energy Range20,000 µE/s/m2
  • Depth (in)1 9/16
  • Height (in)1 13/16
  • Height (cm)4.6
  • Depth (cm)3.96
  • DescriptionVisible PAR Sensor 1-13/16\"H x 1-9/16\" dia
  • Warranty1 year
1 Year WarrantyNIST Traceable Calibration
Key Features
  • 1 year warranty
More About this Item
Replacement sensors for your Solar Light radiometer.
Measures light intensity below 0.01 lux and can also be used as a safety detector for hazardous intensities of light. Measures photosynthetic active radiation.
Weatherproof detector housing available.