Simpson Meter 0-1DCMA 4.5 DSP
Representative image only.
Simpson Mfr # 21628 – Item # WW-68489-67
  • 1 year warranty
₹211,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • 3-1/2 (F35) or 4-1/2 (F45) digit, 0.5" High Red LED Display
  • Front Panel Pops Off For Quick Changes in Decimal Point of Display
  • Scaling
  • 120 VAC Power Supply
  • Application Flexibility - inputs available for DC, AC, or ACTRMS voltage or current, temperature, or frequency
  • 9-32 VDC Power Available (option)
  • Panel Cutout: 1.77" x 3.62" Depth: 3.12"