Simpson 260-8P 12391 Relay Protected Black Analog Multimeter
Representative image only.
Simpson Mfr # 12391 – Item # WW-20006-07
Accuracy for the lab, rugged construction for the field
  • Excellent for adjustments such as nulling and peaking
  • Large color code scale
  • AC frequency response to 100 KHz
  • Zero RFI
  • Standard and high energy overload fusing
  • Varistor protected meter movement
  • Mirrored scale
  • Broad range of accessories and probes
  • Wide range capabilities
  • Overload protection relay circuits
  • 1 year warranty
PRODUCT OPTIONS
₹141,000.00INR / Each
₹20,200.00 - ₹27,000.00INR / Each
for an additional
₹20,200.00INR / Each
for an additional
₹27,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRelay Protected Black Analog Multimeter
  • Warranty1 Year
1 Year Warranty
Key Features
  • Excellent for adjustments such as nulling and peaking
  • Large color code scale
  • AC frequency response to 100 KHz
  • Zero RFI
  • Standard and high energy overload fusing
  • Varistor protected meter movement
  • Mirrored scale
  • Broad range of accessories and probes
  • Wide range capabilities
  • Overload protection relay circuits
  • 1 year warranty
More About this Item
The Simpson 260 analog meter is ideal when monitoring fluctuating trends and rates. It features more attributes than its predecessors. Standard and high energy fusing in conjunction with diode network meter movement protection makes
the meter the best combination of quality and affordability. An additional relay overload protection circuit is resettable and provides audible continuity checking.