Simpson 1357MD 03200 Ac Ammeter W/Iron Vane 0-25 3 1/2in Model:03200
Representative image only.
Simpson Mfr # 1357MD | 03200 – Item # WW-68487-57
SPECS & DESCRIPTION
Specifications & Description
  • Display Type3.5\" analog dial
  • Max DC Current mA (mA)25000
  • Min DC Current mA (mA)0
  • DescriptionAc Ammeter W/Iron Vane 0-25 3 1/2in Model:03200
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis