Simpson 1329A 09990 Voltmeter, 0-500 VDC, 4.5 UL WV
Representative image only.
Simpson Mfr # 09990 – Item # WW-68489-02
₹36,400.00INR / Each
₹20,200.00 - ₹41,400.00INR / Each
for an additional
₹20,200.00INR / Each
for an additional
₹27,000.00INR / Each
for an additional
₹33,400.00INR / Each
for an additional
₹27,000.00INR / Each
for an additional
₹33,700.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Display Type4.5\" analog dial
  • Max DC Voltage VDC (VDC)500
  • Min DC Voltage VDC (VDC)0
  • DescriptionVoltmeter, 0-500 VDC, 4.5 UL WV
More About this Item
FEATURES
  • Clear acrylic window for wide angle visibility
  • Rugged black phenolic case
  • Black spade pointer for easy distant reading
  • UL recognized
Additional ranges available. Contact Davis