Silicone Gasket, 10 MESH, Size 2"
Representative image only.
Item # WW-30523-08
Protect your pump, valves and other components in fluid line
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
PRODUCT OPTIONS
₹11,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPlatinum-cured silicone
  • Connection TypeSanitary
  • Sanitary Gasket Size (in)2
  • Mesh Size10
  • Particle Size Retained1905 microns
  • DescriptionSilicone Gasket, 10 MESH, Size 2"
Key Features
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
More About this Item
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derived ingredient free. Each gasket sold individually.
Platinum-cured silicone gaskets are the ideal choice for pharmaceutical, high-purity water, injectable, food, and beverage applications. Gaskets will resist product adhesion, and are tasteless, odorless and do not contain plasticizers. Use silicone gaskets at temperatures from –58 to 446degF (–50 to 230degC); saniti