SHOWA 558-10 Liquid-Proof Reusable Latex Gloves, Size XL, 18"L; 12 Pairs/Pk
Representative image only.
SHOWA Mfr # 558-10 – Item # WW-86288-53
Superior 50 mil thickness
  • Ideal for sandblasting and electroplating
SPECS & DESCRIPTION
Specifications & Description
  • MaterialLatex
  • Thickness (mil)50
  • SizeX-Large
  • Length (in)18
  • Length (cm)46
  • ColorBlack and Red
  • SterileNo
  • DescriptionLiquid-Proof Reusable Latex Gloves, Size XL, 18\"L; 12 Pairs/Pk
Key Features
  • Ideal for sandblasting and electroplating
More About this Item
HD™ heavy-duty latex gloves feature two-color construction that shows wear—gives added safety for sandblasting and electroplating. Not designed for electrical use.