This high-intensity stroboscope utilizes flashing LED lights in synchronous frequencies to operating machinery, shafts, and gears, allowing you to view slowed or frozen images for easy inspection. The LED technology reduces maintenance and downtime by eliminating bulb replacements found in typical xenon-powered stroboscopes.
The intuitive keypad allows you to easily adjust flash rates to coincide with changing speeds of your equipment and the dial control makes fine tuning adjustments quick and simple. Unit features x2 and ÷2 for large spanning over the entire flash range, and phase shift capability for rotating equipment letting you easily change the focus on blades, gears, or alternate sections of machinery.