Sherwood Scientific 500 350 24 Filter top cap for 5-L sealed tub, 3 um/polyester
Representative image only.
Sherwood Scientific Mfr # 500 350 24 – Item # WW-02658-69
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFilter top cap for 5-L sealed tub, 3 um/polyester
More About this Item
Select cap 02658-68 for wetter or denser samples requiring more air flow. Use filter top cap 02658-69 for drying 5-µm particles.
Note: Only 100 g of sample can be dried at a time with filter top cap 02658-69.