Setra 600751 Bayonet Electrical Connector Adapter, female 6-pin, ASM
Representative image only.
Setra Mfr # 600751 – Item # WW-98075-90
Optimizer performance of your pressure transducer
  • 1 year warranty
PRODUCT OPTIONS
₹17,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrical Connections6-pin micro connector
  • DescriptionBayonet Electrical Connector Adapter, female 6-pin, ASM
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 66 Rated
Key Features
  • 1 year warranty
More About this Item
These accessories are compatible with the Setra 98075-XX series AccuSense™ High-Accuracy Pressure Transducers.
The SecureCal™ calibrator 98075-95 is used to calibrate transducers in the field—enables zero-tare and snap-span adjustment, factory calibration restore function, and two-line LCD/LED feedback.